Isosorbide dimethyl ether 97%

Supplier: Ambeed
Warning

Synonyms: (3R,3aR,6S,6aR)-3,6-Dimethoxyhexahydrofuro[3,2-b]furan

A113999-100G A113999-25G A113999-5G A113999-1000G A113999-500G
76796-972EA 11 USD
76796-972 76796-974 76796-976 76796-978 77570-762 77878-853
Isosorbide dimethyl ether 97%
Isosorbide dimethyl ether

Caution: for Research Use Only

Formula: C₈H₁₄O₄
MDL Number: MFCD00064828
CAS Number: 5306-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR