Ellagic acid 98%

Supplier: Ambeed
Warning

A106247-100G A106247-1G A106247-250MG A106247-25G A106247-500G A106247-5G
76796-534EA 143 USD
76796-534 76796-536 76796-538 76796-540 76796-542 76796-544
Ellagic acid 98%
Ellagic acid

Caution: for Research Use Only

Formula: C₁₄H₆O₈
MDL Number: MFCD00006914
CAS Number: 476-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR