Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride 98%

Supplier: Ambeed
Warning

A879638-100MG A879638-1G A879638-250MG A879638-5G
76792-012EA 12 USD
76792-012 76792-014 76792-016 76792-018
Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride 98%
Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) dichloride

Caution: for Research Use Only

Formula: C₇₂H₄₈Cl₂N₆Ru
MDL Number: MFCD03095387
CAS Number: 36309-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR