Triethylene glycol dimethacrylate 95% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A836977-100G A836977-25G A836977-500G
76791-840EA 20 USD
76791-840 76791-842 76791-844
Triethylene glycol dimethacrylate 95% stabilized
Triethylene glycol dimethacrylate

Caution: for Research Use Only

Formula: C₁₄H₂₂O₆
MDL Number: MFCD00008591
CAS Number: 109-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR