Uric acid 98%

Supplier: Ambeed
Warning

A744554-5G A744554-25G
76791-266EA 28 USD
76791-266 77569-408
Uric acid 98%
Uric acid

Caution: For research use only.

Formula: C₅H₄N₄O₃
MDL Number: MFCD00005712
CAS Number: 69-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR