(E)-(4-Methoxystyryl)boronic acid 98%

Supplier: Ambeed
Warning

A657012-1G A657012-250MG A657012-5G
76790-870EA 100.99 USD
76790-870 76790-872 76790-874
(E)-(4-Methoxystyryl)boronic acid 98%

Caution: for Research Use Only

Formula: C₉H₁₁BO₃
MDL Number: MFCD04039879
CAS Number: 72316-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR