Tetraphenylethylene 97%

Supplier: Ambeed
Warning

A524263-100G A524263-1G A524263-250MG A524263-25G A524263-5G
76790-110EA 224 USD
76790-110 76790-112 76790-114 76790-116 76790-118
Tetraphenylethylene 97%
Tetraphenylethylene

Caution: for Research Use Only

Formula: C₂₆H₂₀
MDL Number: MFCD00004764
CAS Number: 632-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR