Methyl-5-bromo-2-fluoronicotinate 98%

Supplier: Ambeed
Warning

A500404-1G A500404-250MG A500404-5G A500404-10G A500404-25G
76789-918EA 27 USD
76789-918 76789-920 76789-922 77669-246 77872-836
Methyl-5-bromo-2-fluoronicotinate 98%
Methyl-5-bromo-2-fluoronicotinate

Caution: for Research Use Only

Formula: C₇H₅BrFNO₂
MDL Number: MFCD11617316
CAS Number: 931105-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR