Terfenadine 98%

Supplier: Ambeed
Danger

A373035-100MG A373035-1G A373035-1MG A373035-250MG A373035-25MG A373035-5G
76788-704EA 26 USD
76788-704 76788-706 76788-708 76788-710 76788-712 77872-573
Terfenadine 98%
Terfenadine

Caution: for Research Use Only

Formula: C₃₂H₄₁NO₂
MDL Number: MFCD00079622
CAS Number: 50679-08-8
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR