Tin(II) oxide 99%

Supplier: Ambeed
Danger

A273068-25G A273068-100G
76787-222EA 7 USD
76787-222 77872-323
Tin(II) oxide 99%
Tin (II) oxide

Caution: for Research Use Only

Formula: OSn
MDL Number: MFCD00011243
CAS Number: 21651-19-4
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR