Trithiocyanuric acid 98%
Supplier: Ambeed
|
Warning
|
A250986-100G
A250986-10G
A250986-500G
A250986-25G
76786-822EA
34
USD
76786-822
76786-824
76786-826
77304-546
Trithiocyanuric acid 98%
Trithiocyanuric acid
Caution: for Research Use Only
Formula:
C₃H₃N₃S₃ |
MDL Number:
MFCD00006052 CAS Number: 638-16-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...