QNZ 98%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-4-[2-(4-phenoxyphenyl)ethylamino]quinazoline, N4-(4-Phenoxyphenethyl)quinazoline-4,6-diamine

A230954-10MG A230954-1MG A230954-50MG A230954-5MG A230954-100MG
76786-248EA 79 USD
76786-248 76786-250 76786-252 76786-254 77872-174
QNZ 98%
QNZ

Caution: for Research Use Only

Formula: C₂₂H₂₀N₄O
MW: 356.429 g/mol
MDL Number: MFCD06411436
CAS Number: 545380-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR