Thiazovivin ≥98%

Supplier: Ambeed
Warning

Synonyms: N-Benzyl-2-(pyrimidin-4-ylamino)thiazole-4-carboxamide

A211276-10MG A211276-1MG A211276-5MG A211276-250MG A211276-100MG A211276-1G
76785-672EA 47 USD
76785-672 76785-674 76785-676 77600-698 77667-764 77667-766
Thiazovivin ≥98%
Thiazovivin

Caution: for Research Use Only

Formula: C₁₅H₁₃N₅OS
MDL Number: MFCD16495823
CAS Number: 1226056-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR