Tolcapone 98%

Supplier: Ambeed
Danger

Synonyms: 3,4-Dihydroxy-4'-methyl-5-nitrobenzophenone

A207157-100MG A207157-1MG A207157-250MG A207157-50MG A207157-1G
76785-554EA 113 USD
76785-554 76785-556 76785-558 76785-560 77667-740
Tolcapone 98%
Tolcapone

Caution: for Research Use Only

Formula: C₁₄H₁₁NO₅
MW: 273.24 g/mol
MDL Number: MFCD00866569
CAS Number: 134308-13-7
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR