Squalene 98+%
Supplier: Ambeed
|
Danger
|
Synonyms:
2,6,10,15,19,23-Hexamethyltetracosa-2,6,10,14,18,22-hexaene
A206951-100G
A206951-25G
A206951-500G
A206951-5G
76785-532EA
30
USD
76785-532
76785-534
76785-536
76785-538
Squalene 98+%
Squalene
Caution: for Research Use Only
Formula:
C₃₀H₅₀ MW: 410.73 g/mol |
MDL Number:
MFCD00008912 CAS Number: 111-02-4 UN: 2810 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...