Boc-Phe-OH 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-DL-phenylalanine, 2-((tert-Butoxycarbonyl)amino)-3-phenylpropanoic acid, Boc-DL-Phe-OH

A205783-100G A205783-25G A205783-5G
76785-490EA 59 USD
76785-490 76785-492 76785-494
Boc-Phe-OH 98%
Boc-Phe-OH

Caution: for Research Use Only

Formula: C₁₄H₁₉NO₄
MW: 265.309 g/mol
MDL Number: MFCD00558969
CAS Number: 4530-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR