4-(Thiazol-2-yloxy)phenylamine 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Thiazol-2-yloxy)aniline

A196312-100MG A196312-1G A196312-250MG A196312-5G
76785-128EA 22 USD
76785-128 76785-130 76785-132 76785-134
4-(Thiazol-2-yloxy)phenylamine 95%
4-(Thiazol-2-yloxy)phenylamine

Caution: for Research Use Only

Formula: C₉H₈N₂OS
MW: 192.24 g/mol
MDL Number: MFCD09743704
CAS Number: 105350-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR