Sulfasalazine 98%

Supplier: Ambeed
Warning

A176325-100G A176325-10G A176325-1G A176325-25G A176325-500G A176325-5G
76784-456EA 57 USD
76784-456 76784-458 76784-460 76784-462 76784-464 76784-466
Sulfasalazine 98%
Sulfasalazine

Caution: for Research Use Only

Formula: C₁₈H₁₄N₄O₅S
MDL Number: MFCD00057363
CAS Number: 599-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR