(4-Chloro-3-(trifluoromethyl)phenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-3-(trifluoromethyl)phenylacetic acid

A158378-100MG A158378-1G A158378-250MG A158378-5G
76783-718EA 9 USD
76783-718 76783-720 76783-722 76783-724
(4-Chloro-3-(trifluoromethyl)phenyl)acetic acid 98%
(4-Chloro-3-(trifluoromethyl)phenyl)acetic acid

Caution: for Research Use Only

Formula: C₉H₆ClF₃O₂
MW: 238.59
MDL Number: MFCD04116012
CAS Number: 22902-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR