Triethylammonium bromide 95%

Supplier: Ambeed
Warning

Synonyms: Triethylamine hydrobromide

A146386-100G A146386-10G A146386-25G
76783-166EA 43 USD
76783-166 76783-168 77317-076
Triethylammonium bromide 95%
Triethylammonium bromide

Caution: for Research Use Only

Formula: C₆H₁₅N·HBr
MW: 182.1
MDL Number: MFCD00054287
CAS Number: 636-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR