Tris(2,4-di-tert-butylphenyl)phosphite 97% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: Tris(2,4-di-t-butylphenyl)phosphite, Phosphorous Acid Tris(2,4-di-tert-butylphenyl) Ester, Tris(2,4-di-tert-butylphenyl) phosphite

A127499-100G A127499-25G A127499-500G
76782-106EA 6 USD
76782-106 76782-108 76782-110
Tris(2,4-di-tert-butylphenyl)phosphite 97% (by HPLC)
Tris(2,4-di-tert-butylphenyl)phosphite

Caution: for Research Use Only

Formula: C₄₂H₆₃O₃P
MDL Number: MFCD00072706
CAS Number: 31570-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR