ThinkLite ICON Healer Pro Air Purifier
Supplier: Unimed
ThinkLite
TIAH1039PRO
76716-772EA 17680.01 USD
76716-772
ThinkLite ICON Healer Pro Air Purifier
Air Purifiers
Continuously clean any building’s air to the highest health and safety standards.
  • Significantly lessens the transmission of disease
  • Boosts focus and productivity for workers
  • Improves cognitive function
  • Leads to less absenteeism due to illness
  • Reduces the impact of asthma and allergies
  • Supports deep, restful sleep
  • Lowers the risk of respiratory illness
Contaminated air from the space is continuously drawn in through the inlet at the bottom of the unit. The air first passes through the custom designed carbon pre-filter. This filters out larger particles and heavy dust while layers of activated carbon absorb volatile organic compounds (TVOCs).
Next, air enters the neutralization chamber, which is equipped with a patented combination of UVC and UVA light and other technologies, including PCO. Here, all remaining TVOCs are eliminated while particles as small as 0.1 micron, including viruses and other pathogens, are destroyed.
Finally, the purified air is propelled through high-grade ULPA filters to ensure total filtration of the smallest particles, even smaller than 0.1 micron. Each filter has an active carbon layer for extra VOC absorption. Pure, clean air leaves the unit through the heated air outlet on the sides of the ICON M unit.