Sodium-L(+)-glutamate monohydrate 99%

Supplier: Ambeed
Warning

Synonyms: (S)-(+)-2-Aminopentanedioic acid sodium salt monohydrate, Sodium L-Glutamate Monohydrate, L(+)-Glutamic acid sodium salt monohydrate, L(+)-2-Aminopentanedioic acid sodium salt monohydrate, (S)-(+)-Glutamic acid sodium salt monohydrate, L(+)-Glutamic acid monosodium salt monohydrate, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-Sodium glutamate monohydrate, L(+)-α-Glutamic acid sodium salt monohydrate, L(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-α-Glutamic acid sodium salt monohydrate

A749276-1000G A749276-100G A749276-25G A749276-500G
76776-206EA 52 USD
76776-206 76776-208 76776-210 76776-212
Sodium-L(+)-glutamate monohydrate 99%
Sodium-L(+)-glutamate monohydrate

Caution: for Research Use Only

Formula: C₅H₈NNaO₄·H₂O
MW: 187.13 g/mol
MDL Number: MFCD00150138
CAS Number: 6106-04-3
Merck Index: 13,06278

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR