3,5-Difluorobenzyl alcohol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(3,5-Difluorophenyl)methanol
A695070-25G
A695070-5G
A695070-100G
76775-860EA
29
USD
76775-860
76775-862
77878-530
3,5-Difluorobenzyl alcohol 98%
3,5-Difluorobenzyl alcohol
Caution: for Research Use Only
Formula:
C₇H₆F₂O MW: 144.12 g/mol |
MDL Number:
MFCD00010313 CAS Number: 79538-20-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...