2-Isobutylthiazole 98%

Supplier: Ambeed
Danger

A646623-5G A646623-25G
76775-538EA 6.37 USD
76775-538 76775-540 77878-464
2-Isobutylthiazole 98%
2-Isobutylthiazole

Caution: for Research Use Only

Formula: C₇H₁₁NS
MDL Number: MFCD00005334
CAS Number: 18640-74-9
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR