5(6)-TAMRA 98% mix
Supplier: Ambeed
|
Warning
|
Synonyms:
TAMRA 5 and 6-isomer, Tetramethylrhodamine-5-(and-6)-maleimide, 5(6)-Carboxy-tetramethylrhodamine, 9-[2,4(or 2,5)-Dicarboxyphenyl]-3,6-bis(dimethylamino)-xanthylium inner salt
A559208-250MG
A559208-100MG
A559208-1G
76774-906EA
176
USD
76774-906
77594-692
77627-154
5(6)-TAMRA 98% mix
5(6)-TAMRA
Caution: for Research Use Only
Formula:
C₂₅H₂₂N₂O₅ MW: 430.45 g/mol |
MDL Number:
MFCD16661194 CAS Number: 98181-63-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...