N-Boc-amino-(4-N-boc-piperidinyl)carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-4-(Boc-amino)piperidine-4-carboxylic Acid

A524705-25G A524705-1G
76774-676EA 372 USD
76774-676 77627-066
N-Boc-amino-(4-N-boc-piperidinyl)carboxylic acid 98%
N-Boc-amino-(4-N-boc-piperidinyl)carboxylic acid

Caution: for Research Use Only

Formula: C₁₆H₂₈N₂O₆
MW: 344.408 g/mol
MDL Number: MFCD02683116
CAS Number: 189321-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR