Rubrene 99%

Supplier: Ambeed
Warning

Synonyms: 5,6,11,12-Tetraphenylnaphthacene, 5,6,11,12-Tetraphenyltetracene

A345118-100MG A345118-250MG
76773-228EA 10 USD
76773-228 77626-660
Rubrene 99%
Rubrene

Caution: for Research Use Only

Formula: C₄₂H₂₈
MW: 532.68 g/mol
MDL Number: MFCD00003703
CAS Number: 517-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR