Dibenzofuran 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Dibenzo[b,d]furan
A323273-10G
A323273-25G
A323273-500G
A323273-100G
76772-956EA
5
USD
76772-956
76772-958
77593-916
77598-424
Dibenzofuran 98%
Dibenzofuran
Caution: for Research Use Only
Formula:
C₁₂H₈O MW: 168.19 g/mol Boiling Pt: 285 |
MDL Number:
MFCD00004968 CAS Number: 132-64-9 UN: 3077 ADR: 9 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...