(±)-α-Methoxyphenylacetic acid 98%

Supplier: Ambeed
Warning

A313122-100G A313122-1G A313122-25G A313122-500G A313122-5G
76772-802EA 275 USD
76772-802 76772-804 76772-806 76772-808 76772-810
(±)-α-Methoxyphenylacetic acid 98%
(±)-α-Methoxyphenylacetic acid

Caution: for Research Use Only

Formula: C₉H₁₀O₃
MDL Number: MFCD00064217
CAS Number: 7021-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR