3',5'-Dibromo-4'-hydroxyacetophenone 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
1-(3,5-Dibromo-4-hydroxyphenyl)ethanone
A279810-1G
A279810-5G
76772-286EA
14
USD
76772-286
77626-440
3',5'-Dibromo-4'-hydroxyacetophenone 98%
3',5'-Dibromo-4'-hydroxyacetophenone
Caution: for Research Use Only
Formula:
C₈H₆Br₂O₂ MW: 293.9400 g/mol |
MDL Number:
MFCD00075779 CAS Number: 2887-72-1 UN: 1759 ADR: 8 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...