2',3'-Dideoxyadenosine 98%

Supplier: Ambeed
Warning

Synonyms: ((2S,5R)-5-(6-Amino-9H-purin-9-yl)tetrahydrofuran-2-yl)methanol

A204150-1G A204150-250MG
76770-266EA 302.36 USD
76770-266 77598-302
2',3'-Dideoxyadenosine 98%
2',3'-Dideoxyadenosine

Caution: for Research Use Only

Formula: C₁₀H₁₃N₅O₂
MW: 235.24 g/mol
MDL Number: MFCD00010534
CAS Number: 4097-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR