2-Fluorobenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: o-Fluorobenzoic acid

A129612-1000G A129612-100G A129612-10G A129612-25G A129612-500G
76767-132EA 63 USD
76767-132 76767-134 76767-136 76767-138 76767-140
2-Fluorobenzoic acid 98%
2-Fluorobenzoic acid

Caution: for Research Use Only

Formula: C₇H₅FO₂
MDL Number: MFCD00002405
CAS Number: 445-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR