4-Chloro-2-methylphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-2-methylbenzeneboronic acid

A120839-100G A120839-10G A120839-1G A120839-25G A120839-5G
76766-448EA 177 USD
76766-448 76766-450 76766-452 76766-454 77596-264
4-Chloro-2-methylphenylboronic acid 98%
4-Chloro-2-methylphenylboronic acid

Caution: for Research Use Only

Formula: C₇H₈BClO₂
MW: 170.40 g/mol
MDL Number: MFCD02683107
CAS Number: 209919-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR