Sodium hexanoate 99%

Supplier: Ambeed
Warning

Synonyms: N-caproicacidsodiumsalt, Sodium Hexanoate

A834398-25G A834398-100G A834398-5G
76728-762EA 50.93 USD
76728-762 77670-200 77670-202
Sodium hexanoate 99%
Sodium hexanoate

Caution: for Research Use Only

Formula: C₆H₁₁NaO₂
MW: 138.1401 g/mol
MDL Number: MFCD00059056
CAS Number: 10051-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR