Pemirolast potassium 98%

Supplier: Ambeed
Warning

A785978-10MG A785978-1G A785978-250MG A785978-25MG A785978-50MG A785978-5G A785978-1MG
76728-386EA 6 USD
76728-386 76728-388 76728-390 76728-392 76728-394 76728-396 77603-020
Pemirolast potassium 98%
Pemirolast potassium

Caution: for Research Use Only

Formula: C₁₀H₇KN₆O
MDL Number: MFCD01690051
CAS Number: 100299-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR