Pentafluorobenzene 97%

Supplier: Ambeed
Danger

A776900-25G A776900-5G
76728-300EA 19 USD
76728-300 76728-302
Pentafluorobenzene 97%
Pentafluorobenzene

Caution: for Research Use Only

Formula: C₆HF₅
MDL Number: MFCD00000286
CAS Number: 363-72-4
UN: 2924
ADR: 3,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR