N-(1-Pyrenyl)maleimide 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Pyren-1-yl)-1H-pyrrole-2,5-dione

A758786-100MG A758786-1G A758786-250MG A758786-5G
76728-182EA 38 USD
76728-182 76728-184 76728-186 77670-040
N-(1-Pyrenyl)maleimide 98%
N-(1-Pyrenyl)maleimide

Caution: for Research Use Only

Formula: C₂₀H₁₁NO₂
MW: 297.31 g/mol
MDL Number: MFCD00049301
CAS Number: 42189-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR