Nickel(II) oxalate dihydrate 99%

Supplier: Ambeed
Danger

Synonyms: Nickel oxalate dihydrate

A703284-25G A703284-5G
76727-704EA 106 USD
76727-704 76727-706
Nickel(II) oxalate dihydrate 99%
Nickel(II) oxalate dihydrate

Caution: for Research Use Only

Formula: C₂H₄NiO₆
MDL Number: MFCD00167155
CAS Number: 6018-94-6
UN: 3288
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR