Myristyl myristate 95%

Supplier: Ambeed
Warning

Synonyms: Tetradecyl tetradecanoate

A659771-100G A659771-25G A659771-500G
76727-416EA 25 USD
76727-416 76727-418 76727-420
Myristyl myristate 95%
Myristyl myristate

Caution: for Research Use Only

Formula: C₂₈H₅₆O₂
MW: 424.7431 g/mol
MDL Number: MFCD00056201
CAS Number: 3234-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR