Salubrinal 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(E)-N-(2,2,2-Trichloro-1-(3-(quinolin-8-yl)thioureido)ethyl)cinnamamide
A591957-10MG
A591957-1MG
A591957-25MG
A591957-50MG
A591957-5MG
A591957-100MG
A591957-250MG
76726-836EA
41
USD
76726-836
76726-838
76726-840
76726-842
76726-844
77669-556
77872-976
Salubrinal 98%
Salubrinal
Caution: for Research Use Only
Formula:
C₂₁H₁₇Cl₃N₄OS MW: 479.81 g/mol |
MDL Number:
MFCD00548612 CAS Number: 405060-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...