Nickel(II) bromide trihydrate 97% (Ni≥20.5%)

Supplier: Ambeed
Danger

A565841-25G A565841-5G A565841-100G
76726-580EA 12 USD
76726-580 76726-582 77572-800
Nickel(II) bromide trihydrate 97% (Ni≥20.5%)
Nickel (II) bromide trihydrate

Caution: for Research Use Only

Formula: Br₂Ni·3H₂O
MW: 272.54
MDL Number: MFCD00011141
CAS Number: 7789-49-3
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR