Phenyl propargyl ether 98%

Supplier: Ambeed
Warning

Synonyms: (Prop-2-ynyloxy)benzene, (Prop-2-yn-1-yloxy)benzene

A541885-100G A541885-1G A541885-5G A541885-25G
76726-372EA 215 USD
76726-372 76726-374 76726-376 77669-392
Phenyl propargyl ether 98%
Phenyl propargyl ether

Caution: for Research Use Only

Formula: C₉H₈O
MDL Number: MFCD00048107
CAS Number: 13610-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR