Tetradecylphosphonic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1-Tetradecylphosphonic acid, n-Tetradecylphosphonic acid, TDPA, N-tetradecylphosphonicacid

A529374-1000G A529374-100G A529374-25G A529374-500G
76726-266EA 190.97 USD
76726-266 76726-268 76726-270 76726-272
Tetradecylphosphonic acid 98%
Tetradecylphosphonic acid

Caution: for Research Use Only

Formula: C₁₄H₃₁O₃P
MW: 278.37 g/mol
MDL Number: MFCD00015834
CAS Number: 4671-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR