Methyl myristate 98%

Supplier: Ambeed
Warning

Synonyms: Tetradecylic acid methyl ester, Myristic acid methyl ester, Tetradecanoic acid methyl ester, Methyl tetradecanoate

A481038-100G A481038-25G A481038-500G
76725-874EA 6 USD
76725-874 76725-876 76725-878
Methyl myristate 98%
Methyl myristate

Caution: for Research Use Only

Formula: C₁₅H₃₀O₂
MW: 242.4 g/mol
Boiling Pt: 323
Melting Pt: 18
Flash Pt: 140
MDL Number: MFCD00008983
CAS Number: 124-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR