Butyl myristate 98%

Supplier: Ambeed
Warning

A450218-100G A450218-25G A450218-5G
76725-560EA 58 USD
76725-560 76725-562 76725-564
Butyl myristate 98%
Butyl myristate

Caution: for Research Use Only

Formula: C₁₈H₃₆O₂
MDL Number: MFCD00015077
CAS Number: 110-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR