Silver diethyldithiocarbamate 98%

Supplier: Ambeed
Warning

A431348-25G A431348-5G A431348-100G
76725-398EA 44 USD
76725-398 76725-400 77872-689
Silver diethyldithiocarbamate 98%
Silver diethyldithiocarbamate

Caution: for Research Use Only

Formula: (C₂H₅)₂NCSSAg
MDL Number: MFCD00004929
CAS Number: 1470-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR