N,N-Diethylsalicylamide 97%

Supplier: Ambeed
Warning

A397066-100G A397066-1G A397066-25G A397066-5G A397066-10G
76725-052EA 283 USD
76725-052 76725-054 76725-056 76725-058 77879-856
N,N-Diethylsalicylamide 97%
N,N-Diethylsalicylamide

Caution: for Research Use Only

Formula: C₁₁H₁₅NO₂
MDL Number: MFCD00010816
CAS Number: 19311-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR