N,N-Dimethylaminomethylferrocene 98%

Supplier: Ambeed
Warning

Synonyms: Ferrocenylmethyldimethylamine, (Dimethylaminomethyl)ferrocene

A371697-100G A371697-1G
76724-776EA 238 USD
76724-776 76724-778
N,N-Dimethylaminomethylferrocene 98%
N,N-Dimethylaminomethylferrocene

Caution: for Research Use Only

Formula: C₁₃H₁₇FeN
MDL Number: MFCD00001433
CAS Number: 1271-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR