Pyronin G > 55% (dye content) BS

Supplier: Ambeed
Danger

A353427-1G A353427-25G A353427-5G
76724-492EA 39 USD
76724-492 76724-494 76724-496
Pyronin G > 55% (dye content) BS
Pyronine G

Caution: for Research Use Only

Formula: C₁₇H₁₉ClN₂O
MDL Number: MFCD00011725
CAS Number: 92-32-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR